Ethyl-4-hydroxyquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 4-hydroxyquinoline-3-carboxylate

045036-1G 045036-5G
101868-830EA 36.92 USD
101868-830 101868-832
Ethyl-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 4-hydroxyquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MW: 217.2206 g/mol
Melting Pt: 270 °C
MDL Number: MFCD00173406
CAS Number: 26892-90-0

Order Now

Specification Test Results

Melting point 270°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR