Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylate

045034-500MG 045034-1G 045034-5G
101868-816EA 375.38 USD
101868-816 101868-818 101868-820
Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate ≥95%
Ethyl 4-hydroxy-8-methylquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₃
MW: 231.2500 g/mol
Melting Pt: 271-274°C (dec)
MDL Number: MFCD00173390
CAS Number: 77156-75-3

Order Now

Specification Test Results

Melting point 271-274°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR