Ethyl-6-bromo-4-chloroquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

045032-1G 045032-5G
101868-812EA 181.54 USD
101868-812 101868-814
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate ≥95%
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₁₂H₉BrClNO₂
MW: 314.563 g/mol
Boiling Pt: 378 °C
Melting Pt: 100…102 °C
Density: 1.578
Flash Pt: 182 °C
MDL Number: MFCD00173367
CAS Number: 206257-39-8

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR