3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2Z)-3-Amino-3-[4-(3,4-dichlorophenyl)piperazin-1-yl]prop-2-enenitrile

045031-500MG 045031-1G 045031-5G
101868-804EA 747.69 USD
101868-804 101868-806 101868-808
3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile ≥95%
3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile

Hazard: Irritant

Formula: C₁₃H₁₄Cl₂N₄
MW: 297.19 g/mol
Melting Pt: 188…190 °C
MDL Number: MFCD00664795
CAS Number: 1164541-77-8

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR