3-Amino-3-(4-methylpiperazino)acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2E)-3-Amino-3-(4-methylpiperazin-1-yl)prop-2-enenitrile

045030-500MG 045030-1G 045030-5G
101868-798EA 747.69 USD
101868-798 101868-800 101868-802
3-Amino-3-(4-methylpiperazino)acrylonitrile ≥95%
3-Amino-3-(4-methylpiperazino)acrylonitrile

Hazard: Irritant

Formula: C₈H₁₄N₄
MW: 166.23 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00664794
CAS Number: 136062-62-9

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR