[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulfanyl-1H-imidazol-5-yl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl-5-(hydroxymethyl)-1H-imidazole-2-thiol
045028-500MG
045028-1G
101868-788EA
921.54
USD
101868-788
101868-790
[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulfanyl-1H-imidazol-5-yl]methanol ≥95%
[1-(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)-2-sulfanyl-1H-imidazol-5-yl]methanol
Hazard: Irritant
Formula:
C₁₂H₁₂ClF₃N₄OS MW: 352.77 g/mol Melting Pt: 219…221 °C |
MDL Number:
MFCD00215163 CAS Number: 338794-80-2 |
Specification Test Results
Melting point | 219-221°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...