5-(4-Methoxyphenyl)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045027-500MG 045027-1G 045027-5G
101868-782EA 260 USD
101868-782 101868-784 101868-786
5-(4-Methoxyphenyl)-2-thiophenecarboxylic acid ≥95%
5-(4-Methoxyphenyl)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₃S
Melting Pt: 202…204 °C
MDL Number: MFCD00221059
CAS Number: 116016-56-9

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR