Methyl-3-(acetylamino)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-acetamidothiophene-2-carboxylate
045025-500MG
045025-1G
045025-5G
101868-770EA
278.46
USD
101868-770
101868-772
101868-774
Methyl-3-(acetylamino)-2-thiophenecarboxylate ≥95%
Methyl-3-(acetylamino)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₈H₉NO₃S Melting Pt: 94…95 °C |
MDL Number:
MFCD00100044 CAS Number: 22288-79-5 |
Specification Test Results
Melting point | 94-95°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...