3-(Benzoylamino)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045024-500MG 045024-1G 045024-5G
101868-764EA 278.46 USD
101868-764 101868-766 101868-768
3-(Benzoylamino)-2-thiophenecarboxylic acid ≥95%
3-(Benzoylamino)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₉NO₃S
Melting Pt: 200…201 °C
MDL Number: MFCD01316568
CAS Number: 147123-66-8

Order Now

Specification Test Results

Melting point 200-201°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR