7-[2-(2-Thienyl)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

045023-500MG 045023-1G 045023-5G
101868-758EA 747.69 USD
101868-758 101868-760 101868-762
7-[2-(2-Thienyl)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-[2-(2-Thienyl)vinyl][1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₁H₉N₅S
MW: 243.29 g/mol
Melting Pt: 259…261 °C
MDL Number: MFCD00232515
CAS Number: 338793-62-7

Order Now

Specification Test Results

Melting point 259-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR