3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole ≥95%

Supplier: Matrix Scientific

045022-500MG 045022-1G 045022-5G
101868-752EA 556.92 USD
101868-752 101868-754 101868-756
3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole ≥95%
3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₁₁H₉N₅
Melting Pt: 188…190 °C
MDL Number: MFCD00244275
CAS Number: 338793-57-0

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR