Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1-Benzyl-5-oxo-3-pyrrolidinecarboxylate

045018-5G 045018-25G 045018-100G
101868-736EA 30.77 USD
101868-736 101868-738 102663-722
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate ≥95%
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MW: 233.267 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD00003196
CAS Number: 51535-00-3

Order Now

Specification Test Results

Melting point 66-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR