2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

045015-500MG 045015-1G 045015-5G
101868-722EA 135.38 USD
101868-722 101868-724 101868-726
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol ≥95%
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₇H₈F₃N₃O
Melting Pt: 198…200 °C
MDL Number: MFCD00171969
CAS Number: 338792-66-8

Order Now

Specification Test Results

Melting point 198-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR