2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Cyano-3-fluorophenyl)-2-phenylacetonitrile

045008-500MG 045008-1G 045008-5G
101868-692EA 181.54 USD
101868-692 101868-694 101868-696
2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile

Hazard: Irritant

Formula: C₁₅H₉FN₂
MW: 236.25 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD00231973
CAS Number: 338791-66-5

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR