2-[Cyano(phenyl)methyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

045007-500MG 045007-1G 045007-5G
101868-686EA 181.54 USD
101868-686 101868-688 101868-690
2-[Cyano(phenyl)methyl]benzenecarbonitrile ≥95%
2-[Cyano(phenyl)methyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₅H₁₀N₂
Melting Pt: 85…87 °C
MDL Number: MFCD00231972
CAS Number: 127667-03-2

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR