2-[(1-Methyl-3-oxo-3-phenyl-1-propenyl)amino]acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-{[(2E)-4-Oxo-4-phenylbut-2-en-2-yl]amino}acetonitrile

045001-500MG 045001-1G 045001-5G
101868-652EA 181.54 USD
101868-652 101868-654 101868-656
2-[(1-Methyl-3-oxo-3-phenyl-1-propenyl)amino]acetonitrile ≥95%
2-[(1-Methyl-3-oxo-3-phenyl-1-propenyl)amino]acetonitrile

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD00102733
CAS Number: 56464-51-8

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR