Diethyl-4-chloro-2,6-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

045000-500MG 045000-1G
101868-648EA 698.46 USD
101868-648 101868-650
Diethyl-4-chloro-2,6-pyridinedicarboxylate ≥95%
Diethyl-4-chloro-2,6-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₄
Melting Pt: 91…93 °C
MDL Number: MFCD00215521
CAS Number: 53389-01-8

Order Now

Specification Test Results

Melting point 91-93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR