5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine ≥95%

Supplier: Matrix Scientific

044999-500MG 044999-1G 044999-5G
101868-642EA 747.69 USD
101868-642 101868-644 101868-646
5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine ≥95%
5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine

Hazard: Irritant

Formula: C₁₁H₃F₆N₅
Melting Pt: 138…140 °C
MDL Number: MFCD00141799
CAS Number: 338786-45-1

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR