1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

044989-500MG 044989-1G 044989-5G
101868-602EA 710.77 USD
101868-602 101868-604 101868-606
1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-[3,5-Bis(trifluoromethyl)benzyl]-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₅H₉F₆NO₃
Melting Pt: 168…169 °C
MDL Number: MFCD00243697
CAS Number: 338781-55-8

Order Now

Specification Test Results

Melting point 168-169°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR