4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%

Supplier: Matrix Scientific

044987-500MG 044987-1G 044987-5G
101868-590EA 181.54 USD
101868-590 101868-592 101868-594
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂O
MW: 288.66 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD00170933
CAS Number: 71422-80-5

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR