Diethyl-2-{[3-(bromomethyl)-2-thienyl]-methylene}malonate ≥95%

Supplier: Matrix Scientific

044986-500MG 044986-1G 044986-5G
101868-584EA 375.38 USD
101868-584 101868-586 101868-588
Diethyl-2-{[3-(bromomethyl)-2-thienyl]-methylene}malonate ≥95%
Diethyl-2-{[3-(bromomethyl)-2-thienyl]-methylene}malonate

Hazard: Irritant

Formula: C₁₃H₁₅BrO₄S
Melting Pt: 60…62 °C
MDL Number: MFCD00180836
CAS Number: 104085-30-5

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR