2-(6-Chloro-3-pyridazinyl)-2-(2-pyridyl)acetonitrile ≥95%

Supplier: Matrix Scientific

044981-500MG 044981-1G 044981-5G
101868-554EA 181.54 USD
101868-554 101868-556 101868-558
2-(6-Chloro-3-pyridazinyl)-2-(2-pyridyl)acetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-(2-pyridyl)acetonitrile

Hazard: Irritant

Formula: C₁₁H₇ClN₄
Melting Pt: 233-235°C
MDL Number: MFCD00140706
CAS Number: 338779-25-2

Order Now

Specification Test Results

Melting point 233-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR