6-Chloro-5-nitroimidazo[2,1-b][1,3]thiazole ≥95%

Supplier: Matrix Scientific

044980-500MG 044980-1G 044980-5G
101868-548EA 747.69 USD
101868-548 101868-550 101868-552
6-Chloro-5-nitroimidazo[2,1-b][1,3]thiazole ≥95%
6-Chloro-5-nitroimidazo[2,1-b][1,3]thiazole

Hazard: Irritant

Formula: C₅H₂ClN₃O₂S
Melting Pt: 192…193 °C
MDL Number: MFCD00138960
CAS Number: 23576-89-8

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR