Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

044978-500MG 044978-1G 044978-5G
101868-536EA 207.69 USD
101868-536 101868-538 101868-540
Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate ≥95%
Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₁₃N₃O₃S
MW: 243.29 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD00139418
CAS Number: 99967-78-9

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR