3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-propen-1-one
044968-500MG
044968-1G
044968-5G
101868-490EA
375.38
USD
101868-490
101868-492
101868-494
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one ≥95%
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one
Hazard: Irritant
Formula:
C₈H₁₀N₂OS MW: 182.243 g/mol Melting Pt: 108…110 °C |
MDL Number:
MFCD00105071 CAS Number: 191729-27-8 |
Specification Test Results
Melting point | 108-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...