2-(5-Methyl-1H-pyrazol-3-yl)pyrazine ≥95%
Supplier: Matrix Scientific
044967-500MG
044967-1G
044967-5G
101868-484EA
750.77
USD
101868-484
101868-486
101868-488
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine ≥95%
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine
Hazard: Irritant
Formula:
C₈H₈N₄ Melting Pt: 122…123 °C |
MDL Number:
MFCD08056640 CAS Number: 192711-19-6 |
Specification Test Results
Melting point | 122-123°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...