2-(1H-Pyrazol-3-yl)pyrazine ≥95%
Supplier: Matrix Scientific
044966-500MG
044966-1G
044966-5G
101868-478EA
747.69
USD
101868-478
101868-480
101868-482
2-(1H-Pyrazol-3-yl)pyrazine ≥95%
2-(1H-Pyrazol-3-yl)pyrazine
Hazard: Irritant
Formula:
C₇H₆N₄ Melting Pt: 155…157 °C |
MDL Number:
MFCD03618495 CAS Number: 111781-54-5 |
Specification Test Results
Melting point | 155-157°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...