tert-Butyl-4-(4-nitrophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%

Supplier: Matrix Scientific

044965-500MG 044965-1G 044965-5G
101868-472EA 375.38 USD
101868-472 101868-474 101868-476
tert-Butyl-4-(4-nitrophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%
tert-Butyl-4-(4-nitrophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₀N₂O₄S
MW: 360.44 g/mol
Melting Pt: 120 °C
MDL Number: MFCD08056639
CAS Number: 886361-87-1

Order Now

Specification Test Results

Melting point 120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR