3-(Dimethylamino)-1-(2-pyrazinyl)-2-buten-1-one ≥95%

Supplier: Matrix Scientific

044964-500MG 044964-1G 044964-5G
101868-466EA 921.54 USD
101868-466 101868-468 101868-470
3-(Dimethylamino)-1-(2-pyrazinyl)-2-buten-1-one ≥95%
3-(Dimethylamino)-1-(2-pyrazinyl)-2-buten-1-one

Hazard: Irritant

Formula: C₁₀H₁₃N₃O
MW: 191.23 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD08056638
CAS Number: 886361-86-0

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR