3-Amino-6-(trifluoromethyl)-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)-1H-indazol-3-amine

044963-500MG 044963-1G 044963-5G
101868-460EA 240 USD
101868-460 101868-462 101868-464
3-Amino-6-(trifluoromethyl)-1H-indazole ≥95%
3-Amino-6-(trifluoromethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₆F₃N₃
MW: 201.148 g/mol
Melting Pt: 170-172°C
MDL Number: MFCD03426695
CAS Number: 2250-55-7

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR