Methyl-4'-bromo-4-nitro[1,1'-biphenyl]-2-carboxylate ≥95%

Supplier: Matrix Scientific

044962-500MG 044962-1G 044962-5G
101868-454EA 556.92 USD
101868-454 101868-456 101868-458
Methyl-4'-bromo-4-nitro[1,1'-biphenyl]-2-carboxylate ≥95%
Methyl-4'-bromo-4-nitro[1,1'-biphenyl]-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₀BrNO₄
MW: 336.15 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD08056637
CAS Number: 886361-85-9

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR