Ethyl-3-(2-pyridinyl)-1H-pyrazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

044961-500MG 044961-1G
101868-450EA 215.38 USD
101868-450 101868-452
Ethyl-3-(2-pyridinyl)-1H-pyrazole-5-carboxylate ≥95%
Ethyl-3-(2-pyridinyl)-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₂
Melting Pt: 96…98 °C
MDL Number: MFCD12827769
CAS Number: 174139-65-2

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR