4-Amino-2-chloropyrimidine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-chloro-5-pyrimidinecarbonitrile

044959-500MG 044959-1G 044959-5G
101868-438EA 375.38 USD
101868-438 101868-440 101868-442
4-Amino-2-chloropyrimidine-5-carbonitrile ≥95%
4-Amino-2-chloropyrimidine-5-carbonitrile

Hazard: Toxic

Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Melting Pt: 252°C
MDL Number: MFCD00052343
CAS Number: 94741-69-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 252°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR