6-Hydroxy-2-naphthaldehyde ≥95%

Supplier: Matrix Scientific

044958-500MG 044958-1G 044958-5G
101868-432EA 806.15 USD
101868-432 101868-434 101868-436
6-Hydroxy-2-naphthaldehyde ≥95%
6-Hydroxy-2-naphthaldehyde

Hazard: Irritant

Formula: C₁₁H₈O₂
MW: 172.1800 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD03427537
CAS Number: 78119-82-1

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR