1-{3-Fluoro-4-[(3-methylbenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

044955-500MG 044955-1G 044955-5G
101868-420EA 375.38 USD
101868-420 101868-422 101868-424
1-{3-Fluoro-4-[(3-methylbenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{3-Fluoro-4-[(3-methylbenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₆H₁₅FO₂
Melting Pt: 68…70 °C
MDL Number: MFCD06659567
CAS Number: 885949-77-9

Order Now

Specification Test Results

Melting point 68-70°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR