(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone ≥95%

Supplier: Matrix Scientific

044953-500MG 044953-1G 044953-5G
101868-410EA 375.38 USD
101868-410 101868-412 101868-414
(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone ≥95%
(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone

Hazard: Irritant

Formula: C₁₈H₁₈F₃N₃O
MW: 349.36 g/mol
Melting Pt: 151 °C
MDL Number: MFCD06659566
CAS Number: 885949-76-8

Order Now

Specification Test Results

Melting point 151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR