1-{2-Hydroxy-4-[(4-methylbenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

044952-500MG 044952-1G 044952-5G
101868-404EA 132.31 USD
101868-404 101868-406 101868-408
1-{2-Hydroxy-4-[(4-methylbenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{2-Hydroxy-4-[(4-methylbenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₆H₁₆O₃
Melting Pt: 96…99 °C
MDL Number: MFCD06659565
CAS Number: 885949-75-7

Order Now

Specification Test Results

Melting point 96-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR