2-[(4-Nitrophenyl)sulfanyl]acetic acid ≥95%

Supplier: Matrix Scientific

044951-500MG 044951-1G 044951-5G
101868-398EA 181.54 USD
101868-398 101868-400 101868-402
2-[(4-Nitrophenyl)sulfanyl]acetic acid ≥95%
2-[(4-Nitrophenyl)sulfanyl]acetic acid

Hazard: Irritant

Formula: C₈H₇NO₄S
Melting Pt: 155…157 °C
MDL Number: MFCD00024709
CAS Number: 3406-75-5

Order Now

Specification Test Results

Melting point 155-157°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR