5-Amino-3-(4-bromophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)-1H-pyrazol-5-amine

044950-500MG 044950-1G 044950-5G
101868-392EA 278.46 USD
101868-392 101868-394 101868-396
5-Amino-3-(4-bromophenyl)-1H-pyrazole ≥95%
5-Amino-3-(4-bromophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₈BrN₃
MW: 238.09 g/mol
Melting Pt: 172-174°C
MDL Number: MFCD00558911
CAS Number: 78583-82-1

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR