Methyl-2-(5-formyl-2-methoxyphenoxy)-2-phenylacetate ≥95%

Supplier: Matrix Scientific

044948-500MG 044948-1G 044948-5G
101868-380EA 278.46 USD
101868-380 101868-382 101868-384
Methyl-2-(5-formyl-2-methoxyphenoxy)-2-phenylacetate ≥95%
Methyl-2-(5-formyl-2-methoxyphenoxy)-2-phenylacetate

Hazard: Irritant

Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
Melting Pt: 77…79 °C
MDL Number: MFCD06411626
CAS Number: 885949-40-6

Order Now

Specification Test Results

Melting point 77-79°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR