4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-N-(3-ethynylphenyl)benzamide

044946-500MG 044946-1G 044946-5G
101868-368EA 181.54 USD
101868-368 101868-370 101868-372
4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide ≥95%
4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₅H₁₀ClNO
MW: 255.71 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00176067
CAS Number: 883017-38-7

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR