2,4-Dichloro-1-(2-propynyloxy)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-1-(prop-2-yn-1-yloxy)benzene

044945-500MG 044945-1G 044945-5G
101868-362EA 181.54 USD
101868-362 101868-364 101868-366
2,4-Dichloro-1-(2-propynyloxy)benzene ≥95%
2,4-Dichloro-1-(2-propynyloxy)benzene

Hazard: Irritant

Formula: C₉H₆Cl₂O
MW: 201.05 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD02311548
CAS Number: 17061-90-4

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR