2-(4-Formylphenoxy)-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

044944-500MG 044944-1G 044944-5G
101868-356EA 375.38 USD
101868-356 101868-358 101868-360
2-(4-Formylphenoxy)-2-phenylacetic acid ≥95%
2-(4-Formylphenoxy)-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₅H₁₂O₄
Melting Pt: 157-159°C
CAS Number: 480994-56-7

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR