2-Bromo-4-methoxyphenol ≥97%
Supplier: Matrix Scientific
044943-5G
044943-25G
101868-352EA
92.31
USD
101868-352
101868-354
2-Bromo-4-methoxyphenol ≥97%
2-Bromo-4-methoxyphenol
Hazard: Irritant
Formula:
C₇H₇BrO₂ Melting Pt: 47 °C |
MDL Number:
MFCD02113712 CAS Number: 17332-11-5 |
Specification Test Results
Melting point | 47°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...