2-[(2-Chloro-4-nitrophenyl)sulfanyl]acetic acid ≥95%

Supplier: Matrix Scientific

044942-500MG 044942-1G 044942-5G
101868-346EA 181.54 USD
101868-346 101868-348 101868-350
2-[(2-Chloro-4-nitrophenyl)sulfanyl]acetic acid ≥95%
2-[(2-Chloro-4-nitrophenyl)sulfanyl]acetic acid

Hazard: Irritant

Formula: C₈H₆ClNO₄S
Melting Pt: 108…110 °C
MDL Number: MFCD06411624
CAS Number: 30880-64-9

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR