Methyl-2-(4-acetylphenoxy)-2-phenylacetate ≥95%

Supplier: Matrix Scientific

044940-500MG 044940-1G 044940-5G
101868-334EA 181.54 USD
101868-334 101868-336 101868-338
Methyl-2-(4-acetylphenoxy)-2-phenylacetate ≥95%
Methyl-2-(4-acetylphenoxy)-2-phenylacetate

Hazard: Irritant

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Melting Pt: 100…101 °C
MDL Number: MFCD06200978
CAS Number: 885949-39-3

Order Now

Specification Test Results

Melting point 100-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR