6-(1H-Pyrrol-1-yl)nicotinic acid ≥95%

Supplier: Matrix Scientific

044937-500MG 044937-1G 044937-5G
101868-316EA 355.38 USD
101868-316 101868-318 101868-320
6-(1H-Pyrrol-1-yl)nicotinic acid ≥95%
6-(1H-Pyrrol-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
Melting Pt: 289…291 °C
MDL Number: MFCD00067843
CAS Number: 175135-86-1

Order Now

Specification Test Results

Melting point 289-291°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR