Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
044933-500MG
044933-1G
044933-5G
101868-292EA
921.54
USD
101868-292
101868-294
101868-296
Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate ≥95%
Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₇H₅ClF₃NO₂S MW: 259.64 g/mol Melting Pt: 57…59 °C |
MDL Number:
MFCD02956778 CAS Number: 72850-52-3 |
Specification Test Results
Melting point | 57-59°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...