Methyl-2-amino-6-isopropyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrate ≥95%

Supplier: Matrix Scientific

044932-500MG 044932-1G 044932-5G
101868-286EA 153.85 USD
101868-286 101868-288 101868-290
Methyl-2-amino-6-isopropyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrate ≥95%
Methyl-2-amino-6-isopropyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate hydrate

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃S
MW: 272.37 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD06411621
CAS Number: 1049734-91-9

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR