4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[4-(4-Aminobenzoyl)piperazin-1-yl]benzonitrile

044930-500MG 044930-1G 044930-5G
101868-280EA 375.38 USD
101868-280 101868-282 101868-284
4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile ≥95%
4-[4-(4-Aminobenzoyl)piperazino]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₈H₁₈N₄O
MW: 306.37 g/mol
Melting Pt: 195 °C
MDL Number: MFCD06659560
CAS Number: 885949-73-5

Order Now

Specification Test Results

Melting point 195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR